missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Monoclonal Antibody (OTI9D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813219
This item is not returnable.
View return policy
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Flow CytometrySpecifications
| ICOS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6W8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29851 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry | |
| OTI9D2 | |
| Unconjugated | |
| ICOS | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| Full length human recombinant protein of human ICOS (NP_036224) produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction