Learn More
ICH/USP Oral Target Elements Standard CZ: Ir, Pd, Pt, Rh, Ru @ 100 μg/mL in 5% HCl, VHG™
Description
ICH/USP Oral Target Elements Standard CZ: Ir, Pd, Pt, Rh, Ru @ 100 μg/mL in 5% HCl
Specifications
Specifications
| Chemical Name or Material | Iridium/Palladium/Platinum/Rhodium/Ruthenium |
| CAS | 7439-88-5, 5-3-7440, 6-4-7440, 7440-16-6, 7440-18-8 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Iridium/Palladium/Platinum/Rhodium/Ruthenium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HCl |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Iridium 100 μg/mL/Palladium 100 μg/mL/Platinum 100 μg/mL/Rhodium 100 μg/mL/Ruthenium 100 μg/mL |