Learn More
BINDER™ ICH Light Module with LQC
Product Code. 30238201 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO 720, KBF/KBF PRO 720
KBF/KBF PRO 260/470, KB PRO 260/470
Unit Size:
Each
Product Code. 30238201
Brand: BINDER™ 80122444
Only null left
Add to basket
Add to basket
ICH-Q1B light module with light-dose control
Module consists of:
  • VIS cassette
  • UVA cassette
  • Spherical VIA sensor
  • Spherical UVA sensor
  • Independent power supply
Specifications
For Use With (Equipment) KB PRO 720, KBF/KBF PRO 720
Material Glass
Type Lighting
Includes LQC