Description
- Suitable for ice, wax and other similar materials
- 316 Stainless Steel
- Rotate the sampler as it is pushed into the product
- The sharp edge will cut into the product and the sample will be deposited in the tube
- Push the discharge rod down the Ice Borer to remove the sample
Specifications
Specifications
| Product Type | Ice Borer |
| Material | Stainless Steel (316) |
| Autoclavable | Autoclavable |
| For Use With (Application) | Ice, wax, and other similar materials |
| Dimensions (L x W) | 1000 x 25 mm |
| Length (Metric) | 1000 mm |
| Width (Metric) | 25 mm |