missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27322-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody got 55-80 kDa in western blotting maybe due to glycosylation.
The protein encoded by this gene contains an N-terminal forkhead domain, two BRCA1 C-terminal (BRCT) motifs and a central domain with 13 repetitions of an approximately 41-amino acid sequence. The encoded protein is required to activate the intra-S phase and G2/M phase cell cycle checkpoints in response to DNA damage. This nuclear protein interacts with phosphorylated histone H2AX near sites of DNA double-strand breaks through its BRCT motifs, and facilitates recruitment of the ATM kinase and meiotic recombination 11 protein complex to DNA damage foci.Specifications
ICAM2 | |
Polyclonal | |
Unconjugated | |
ICAM2 | |
CD102, ICAM 2, ICAM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15896, 3384 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13598, P35330 | |
ICAM2 | |
ICAM2 Fusion Protein Ag25952 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |