Learn More
(±)-Ibuprofen Methyl Ester, TRC
Product Code. 30137210
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137210 10 g 10g
30004101 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137210 Supplier Toronto Research Chemicals Supplier No. TRCI14008510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61566-34-5
Molecular Formula C14 H20 O2
Molecular Weight (g/mol) 220.31
Synonym Ibuprofen Methyl Ester
IUPAC Name methyl 2-[4-(2-methylpropyl)phenyl]propanoate
SMILES COC(=O)C(C)c1ccc(CC(C)C)cc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C14H20O2/c1-10(2)9-12-5-7-13(8-6-12)11(3)14(15)16-4/h5-8,10-11H,9H2,1-4H3
Formula Weight 220.1463
Percent Purity >95%
Chemical Name or Material Ibuprofen Methyl Ester
Grade HPLC