Learn More
Ibuprofen Methyl Ester, Mikromol™
Product Code. 30051727
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 30051727
Brand: Mikromol MM0002.53
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ibuprofen Methyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ibuprofen Methyl Ester
CAS 61566-34-5
Analyte or Component Names Ibuprofen Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C14 H20 O2
Certifications/Compliance ISO 17025
Synonym Ibuprofen Methyl Ester
InChI Formula InChI=1S/C14H20O2/c1-10(2)9-12-5-7-13(8-6-12)11(3)14(15)16-4/h5-8,10-11H,9H2,1-4H3
SMILES COC(=O)C(C)c1ccc(CC(C)C)cc1
IUPAC Name methyl 2-[4-(2-methylpropyl)phenyl]propanoate
Molecular Weight (g/mol) 220.31
Quantity 100 mg
Formula Weight 220.1463 g/mol