missing translation for 'onlineSavingsMsg'
Learn More

Ibrutinib Racemate, MedChemExpress

Product Code. p-200099776
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16484338 10 mg 10mg
16455256 100 mg 100mg
16447057 5 mg 5mg
16440667 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16484338

Brand: Medchem Express HY10997A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibrutinib Racemate (PCI-32765 Racemate) is the racemate of Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor with IC50 value of 0.5 nM.

Ibrutinib Racemate (PCI-32765 Racemate) is the racemate of Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor with IC50 value of 0.5 nM.

Specifications

Chemical Name or Material Ibrutinib Racemate
CAS 936563-87-0
Color White
Purity Grade Notes Research
Molecular Formula C25H24N6O2
Quantity 10 mg
Synonym PCI-32765 Racemate
Solubility Information DMSO : 25 mg/mL (56.75 mM; Need ultrasonic)
SMILES C=CC(N1CC(N2N=C(C3=CC=C(OC4=CC=CC=C4)C=C3)C5=C(N)N=CN=C52)CCC1)=O
Molecular Weight (g/mol) 440.5
Formula Weight 440.5
Percent Purity 95.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.