Learn More
Ibrutinib Racemate, MedChemExpress
Product Code. 16484338
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16484338 10 mg 10mg
16455256 100 mg 100mg
16447057 5 mg 5mg
16440667 50 mg 50mg
4 options
Product Code. 16484338
Brand: Medchem Express HY10997A
MedChemExpress Ibrutinib Racemate (PCI-32765 Racemate) is the racemate of Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor with IC50 value of 0.5 nM.
Ibrutinib Racemate (PCI-32765 Racemate) is the racemate of Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor with IC50 value of 0.5 nM.
Specifications
Chemical Name or Material Ibrutinib Racemate
CAS 936563-87-0
Color White
Purity Grade Notes Research
Molecular Formula C25H24N6O2
Quantity 10 mg
Synonym PCI-32765 Racemate
Solubility Information DMSO : 25 mg/mL (56.75 mM; Need ultrasonic)
SMILES C=CC(N1CC(N2N=C(C3=CC=C(OC4=CC=CC=C4)C=C3)C5=C(N)N=CN=C52)CCC1)=O
Molecular Weight (g/mol) 440.5
Formula Weight 440.5
Percent Purity 95.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease