Learn More
Invitrogen™ iBright™ System Contrast Tray
Product Code. 17886511
Change view
Click to view available options
Quantity:
1 Tray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17886511 1 Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17886511 Supplier Invitrogen™ Supplier No. A56597
Estimated Shipment: 10-11-2026
This iBright System Contrast Tray is an iBright imaging system accessory designed to improve the imaging of visible protein gels.
  • This iBright System Contrast Tray is an iBright imaging system accessory designed to improve the imaging of visible protein gels
  • The dual layer tray is designed to create optimal lighting conditions when used with the green LED transilluminator on the iBright imaging system, improving the contrast and lowering the overall background to capture high-quality visible-stain images
  • Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
  • Provides a contrasting background when imaging protein gels stained with colorimetric stains, such as Coomassie blue and silver stains
  • This tray also improves conditions when samples contain dyes that auto-fluoresce under green LED light
  • Recommended for transillumination of crystal violet-stained colony plates
  • A replacement for the White Transilluminator Screen (Cat. No. A33828)
TRUSTED_SUSTAINABILITY
Specifications
Product Line iBright
For Use With (Application) Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Product Type Contrast Tray
Quantity 1 Tray
Shipping Condition Room Temperature