Learn More
Invitrogen™ iBind™ Flex Mini Insert Replacement
Description
The iBind™ Flex Western System offers automated western blot processing. Using customized well inserts, it can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
Specifications
Specifications
| For Use With (Equipment) | iBind |
| Product Line | iBind |
| Type | Replacement Insert |
| Includes | Two wells on row 1 and 3 |
| Quantity | 1 insert |
| Shipping Condition | Room Temperature |