Learn More
Invitrogen™ iBind™ Flex Cards
Product Code. 15857696
Change view
Click to view available options
Type:
Card
Quantity:
10 Cards
40 Cards (10 x 4)
Unit Size:
Pack of 10
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
15857696 Card 40 Cards (10 x 4) Pack of 40
15367238 Card 10 Cards Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15857696 Supplier Invitrogen™ Supplier No. SLF2010X4
Edge
Card for use with iBind™ Flex Western Blot Device.
  • Glass fiber backed card with filer paper stack at base
  • Quantity:  40 Cards (4 boxes of 10)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Chromogenic, Chemiluminescence, Fluorescence
For Use With (Application) Western Blot
For Use With (Equipment) iBind Flex Western Device
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind Flex
Running Time ∼2.5 hr.
Throughput 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card
Type Card
Quantity 40 Cards (10 x 4)
Shipping Condition Room Temperature