Description
IBC discharge valve with innovative hose screw connection. The hose screw connection allows you to attach a wide range of hoses quickly, easily and reliably. No additional hose clamp is needed. Hose fitting PP/POM, external dia. 25 mm.
- 1” flow
- Suitable for connection threads S 60x6 external or 1” external
- Discharge valve and sealing cap PP fibreglass-reinforced, ball and adapter PP
- Max. operating pressure 10 bar (at +20 °C)
- Max. operating temperature 80 °C
Specifications
Specifications
| Material | Polypropylene (Fiber Glass Reinforced) |
| For Use With (Equipment) | IBC Container |
| Includes | Hose screw connection |
| Connectors | Suitable for connection threads S60 x 60 or 1 in. external |