missing translation for 'onlineSavingsMsg'
Learn More

iASPP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18590-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898544

  • 1611.84 NOK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody could both recognize unphosphorylated and phosphorylated iASPP.

This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter′s anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.
Specifications

Specifications

iASPP
Polyclonal
Unconjugated
PPP1R13L
IASPP, Inhibitor of ASPP protein, NKIP1, PPP1R13B like protein, PPP1R13BL, PPP1R13L, Protein iASPP, RAI, RelA associated inhibitor
Rabbit
Antigen Affinity Chromatography
RUO
10848, 333654, 686781
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5I1X5, Q8WUF5
Ppp1r13l
iASPP Fusion Protein Ag13273
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers