Learn More
Invitrogen™ Silencer™ siRNA Construction Kit with manual
Product Code. 10085604
Change view
Click to view available options
Includes:
Kit only
Kit with manual
Unit Size:
Each
Product Code. Includes unitSize
10085604 Kit with manual Each
10299094 Kit only Each
2 options
Product Code. 10085604
Brand: Invitrogen™ AM1620M
The Invitrogen™ Silencer™ siRNA Construction Kit is for the synthesis of siRNAs by in vitro transcription of DNA templates and costs less per siRNA than chemical synthesis. The kit includes sufficient reagents to synthesize 15 siRNAs.
  • Cut siRNA preparation time—generate up to 15 siRNAs in less than 24 hours
  • Get hundreds of transfections per reaction
  • TRUSTED_SUSTAINABILITY
    Specifications
    Content And Storage DNA Hybridization Buffer, T7 Promoter Primer, Klenow Reaction Buffer, 10X dNTP, Exo– Klenow, T7 Enzyme Mix, T7 Reaction Buffer, 2X NTP Mix, Digestion Buffer, DNase, RNase, Sense Control DNA, and Antisense Control DNA should be stored at –20°C. siRNA Binding Buffer, siRNA Wash Buffer, Filter Cartridges, and Tubes should be stored at room temperature. Nuclease-free Water may be stored at any temperature.
    Control Type Transcription Control
    Sample Type In vitro Transcribed siRNA
    Includes Kit with manual
    No. of Reactions 15
    Product Line Silencer, Ambion
    Quantity 15 reactions