Learn More
IKA™ I-MAG SP Set-Up Frame
Product Code. 17553636 Shop All IKA Products
Click to view available options
Model:
I-MAG SP 150
I-MAG SP 300
Capacity (Metric):
200 kg
400 kg
Dimensions (L x W x H):
540 x 500 x 80 mm
740 x 700 x 80 mm
Unit Size:
Each
Product Code. 17553636
Brand: IKA™ 0020102996
Only null left
Add to basket
Add to basket
Stainless steel set-up frame to extend the footprint and load capacity of the I-MAG industrial magnetic stirrer.
  • Set-up Frame
  • Extend the Footprint and Load Capacity of Magnetic Stirrer
  • Accessory for the I-MAG Industrial Magnetic Stirrer
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 400 kg
For Use With (Equipment) I-MAG Industrial Magnetic Stirrer
For Use With (Application) Extend The Footprint and Load Capacity of Magnetic Stirrer
Model I-MAG SP 300
Type Set-Up Frame
Weight (Metric) 20 kg
Dimensions (L x W x H) 740 x 700 x 80 mm
Material Stainless Steel (1.4301)