Description
- Member of the uvex i-range - versatile 3-in-1 safety eyewear range – allows eyewear to be adapted according to workplace hazards
- Large, flat lens delivers unrestricted field of vision
- X-tended sideshield offers additional protection from hazards
- Soft, flexible face seal adapts to the wearer providing both protection and comfort. It also protects against droplets or splashes of liquids and large dust particles (markings 3 and 4)
- Side arms with multi-incline points allow for a high level of individualization to ensure a good fit for all facial shapes and head widths
- Ergonomically shaped side arms provide a secure fit without pressure
- Robust polycarbonate lens with uvex’s proven supravision coating technology – permanent anti-fog and scratch resistant lens
- UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
- Metal-free
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Color | Anthracite, Lime |
| Certifications/Compliance (Europe) | EN 166, EN 170 |
| Lens Tint | Gray |
| Product Type | Ergonomic Safety Glasses |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| UV Protection | UV400 |
| Show More |