missing translation for 'onlineSavingsMsg'
Learn More

I-BRD9, MedChemExpress

Product Code. p-200097132
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487566

Brand: Medchem Express HY18975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress I-BRD9 is a selective cellular chemical probe of bromodomain-containing protein 9 (BRD9) with pIC50 value of 7.3 μM. I-BRD9 has high selectivity for bromodomain and extra terminal domain (BET) family and highly homologous bromodomain-containing protein 7 (BRD7). I-BRD9 can be used to identify genes regulated by BRD9 in Kasumi-1 cells involved in oncology and immune response pathways.

I-BRD9 is a selective cellular chemical probe of bromodomain-containing protein 9 (BRD9) with pIC50 value of 7.3 μM. I-BRD9 has high selectivity for bromodomain and extra terminal domain (BET) family and highly homologous bromodomain-containing protein 7 (BRD7). I-BRD9 can be used to identify genes regulated by BRD9 in Kasumi-1 cells involved in oncology and immune response pathways.

Specifications

Chemical Name or Material I-BRD9
Physical Form Solid
CAS 1714146-59-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H22F3N3O3S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 13.89 mg/mL (27.92 mM; Need ultrasonic)
SMILES N=C(C(S1)=CC2=C1C(C3=CC=CC(C(F)(F)F)=C3)=CN(CC)C2=O)NC(CC4)CCS4(=O)=O
Molecular Weight (g/mol) 497.55
Formula Weight 497.55
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.