missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies HypoThermosol™ FRS

Animal component-free, defined hypothermic (2 - 8°C) preservation medium for a range of cell and tissue types

Brand:  STEMCELL Technologies 07945

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 17109931

  • 21751.70 NOK / 500mL
Estimated Shipment: 03-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • HypoThermosol™ FRS is an optimized hypothermic (2 - 8°C) preservation medium that enables improved and extended preservation of cells, tissues, and organs
  • HypoThermosol™ FRS is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process
  • HypoThermosol™ FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures
  • Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilizers
Specifications

Specifications

Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells
Cryopreservation
500 mL Bag
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers