Learn More
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Product Code. 15361547
Click to view available options
Needle Gauge:
18 G
19 G
20 G
21 G
22 G
23 G
24 G
25 G
26 G
27 G
30 G
Color:
Black
Blue
Brown
Gray
Green
Ivory
Maroon
Orange
Pink
Purple
Yellow
Needle Point Style:
11° Regular
11° Triple Bevel
17° Short
Length (Metric):
13 mm
16 mm
19 mm
23 mm
25 mm
32 mm
38 mm
50 mm
Unit Size:
Pack of 100
Product Code. 15361547
Brand: Terumo™ 8AN2425R1
Make procedures as painless as possible with extremely sharp and reliable stainless steel needles. Terumo™ Agani™ Single-use Sterile Hypodermic Needles feature a unique, latex-free double bevel design for maximum comfort.
  • Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
  • All needles are supplied sterile wrapped in individual, tamper proof peel pouches
  • Simple color coded system allows easy identification of required sizes
  • Use with a wide range of Sterile Syringes
  • Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
  • CE marked and latex free
  • Supplied in boxes of 100 needles
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Class IIa, sterile product following MDD 93/42/EEC
Material Dimethyl Silicone Oil, Epoxy Resin, Polypropylene, Stainless Steel
Color Purple
Needle Gauge 24 G
Diameter (Metric) Outer 0.55 mm
Latex Free Yes
Length (Metric) 25 mm
Product Type Sterile Hypodermic Needle for Single-Use
Disposable Single-use
Quantity Box of 100
Needle Point Style 11° Regular
For Use With (Equipment) Sterile Syringes