Learn More
- Home
- Products
- First Aid and Medical
- Patient Care Products
- Medical Syringes
- Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Product Code. 15331557
Change view
Click to view available options
Needle Gauge:
18 G
19 G
20 G
21 G
22 G
23 G
24 G
25 G
26 G
27 G
30 G
Color:
Black
Blue
Brown
Gray
Green
Ivory
Maroon
Orange
Pink
Purple
Yellow
Needle Point Style:
11° Regular
11° Triple Bevel
17° Short
Length (Metric):
13 mm
16 mm
19 mm
23 mm
25 mm
32 mm
38 mm
50 mm
Unit Size:
Pack of 100
25 product options available for selection
| Product Code. | Needle Gauge | Color | Needle Point Style | Length (Metric) | unitSize |
|---|---|---|---|---|---|
| 15331557 | 27 G | Gray | 11° Regular | 16 mm | Pack of 100 |
| 15351557 | 18 G | Pink | 17° Short | 38 mm | Pack of 100 |
| 15331537 | 19 G | Ivory | 11° Regular | 25 mm | Pack of 100 |
| 15341537 | 19 G | Ivory | 11° Regular | 38 mm | Pack of 100 |
| 15361557 | 19 G | Ivory | 17° Short | 38 mm | Pack of 100 |
| 15428652 | 19 G | Ivory | 11° Regular | 50 mm | Pack of 100 |
| 15371537 | 21 G | Green | 11° Regular | 16 mm | Pack of 100 |
| 15438652 | 21 G | Green | 11° Regular | 50 mm | Pack of 100 |
| 15301547 | 22 G | Black | 11° Regular | 32 mm | Pack of 100 |
| 15331547 | 23 G | Blue | 11° Regular | 16 mm | Pack of 100 |
| 15351547 | 23 G | Blue | 11° Regular | 32 mm | Pack of 100 |
| 15361547 | 24 G | Purple | 11° Regular | 25 mm | Pack of 100 |
| 15391547 | 25 G | Orange | 11° Regular | 25 mm | Pack of 100 |
| 15391557 | 30 G | Yellow | 11° Regular | 13 mm | Pack of 100 |
| 15311557 | 26 G | Brown | 11° Regular | 23 mm | Pack of 100 |
| 15301557 | 26 G | Maroon | 11° Triple Bevel | 13 mm | Pack of 100 |
| 15371547 | 25 G | Orange | 11° Triple Bevel | 16 mm | Pack of 100 |
| 15361537 | 20 G | Yellow | 11° Triple Bevel | 38 mm | Pack of 100 |
| 15321547 | 22 G | Black | 11° Triple Bevel | 38 mm | Pack of 100 |
| 15341547 | 23 G | Blue | 11° Triple Bevel | 25 mm | Pack of 100 |
| 15341557 | 27 G | Gray | 11° Triple Bevel | 19 mm | Pack of 100 |
| 15391537 | 21 G | Green | 11° Triple Bevel | 38 mm | Pack of 100 |
| 15381537 | 21 G | Green | 11° Regular | 25 mm | Pack of 100 |
| 15351537 | 20 G | Yellow | 11° Regular | 25 mm | Pack of 100 |
| 15321537 | 18 G | Pink | 11° Regular | 38 mm | Pack of 100 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
- All needles are supplied sterile wrapped in individual, tamper proof peel pouches
- Simple color coded system allows easy identification of required sizes
- Use with a wide range of Sterile Syringes
- Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
- CE marked and latex free
- Supplied in boxes of 100 needles
Specifications
Specifications
| Certifications/Compliance | Class IIa, sterile product following MDD 93/42/EEC |
| Material | Dimethyl Silicone Oil, Epoxy Resin, Polypropylene, Stainless Steel |
| Color | Gray |
| Needle Gauge | 27 G |
| Diameter (Metric) Outer | 0.4 mm |
| Latex Free | Yes |
| Length (Metric) | 16 mm |
| Product Type | Sterile Hypodermic Needle for Single-Use |
| Disposable | Single-use |
| Quantity | Box of 100 |
| Needle Point Style | 11° Regular |
| For Use With (Equipment) | Sterile Syringes |
| Sterility | Sterile |
| Show More Show Less |