Learn More
BD Hypodermic Needle
Specifications
Specifications
| Color | Yellow |
| Needle Gauge | 30 G |
| Product Type | Hypodermic Needle |
| For Use With (Equipment) | Luer Lock Syringe |
| Sterility | Sterile |