Learn More
Thermo Scientific™ Hypersil™ PREP HS C18 HPLC Columns
Product Code. p-7160182 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
8 mm
20 mm
30 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4.6 mm
10 mm
20 mm
25 mm
30 mm
50 mm
Particle Size:
3 μm
5 μm
10 μm
Column Format:
Guard 1/16 in.
Guard 1/8 in.
Guard Column
Preparative Column, Standard
Preparative Column, Xtended Life
Unit Size:
Each
63 product options available for selection
Product selection table with 63 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Column Format unitSize
16498403 50 mm 2 mm 3 μm Preparative Column, Standard Each
16408413 50 mm 4.6 mm 3 μm Preparative Column, Standard Each
16418413 100 mm 2 mm 3 μm Preparative Column, Standard Each
16428413 100 mm 4.6 mm 3 μm Preparative Column, Standard Each
16438413 150 mm 2 mm 3 μm Preparative Column, Standard Each
16448413 150 mm 4.6 mm 3 μm Preparative Column, Standard Each
16458413 250 mm 2 mm 3 μm Preparative Column, Standard Each
16468413 250 mm 4.6 mm 3 μm Preparative Column, Standard Each
16478413 50 mm 2 mm 5 μm Preparative Column, Standard Each
16488413 50 mm 4.6 mm 5 μm Preparative Column, Standard Each
16498413 50 mm 10 mm 5 μm Preparative Column, Standard Each
16408423 50 mm 20 mm 5 μm Preparative Column, Standard Each
16418423 50 mm 25 mm 5 μm Preparative Column, Xtended Life Each
16428423 100 mm 2 mm 5 μm Preparative Column, Standard Each
16438423 100 mm 4.6 mm 5 μm Preparative Column, Standard Each
16448423 100 mm 10 mm 5 μm Preparative Column, Standard Each
16458423 100 mm 20 mm 5 μm Preparative Column, Standard Each
16468423 100 mm 25 mm 5 μm Preparative Column, Xtended Life Each
16478423 150 mm 2 mm 5 μm Preparative Column, Standard Each
16488423 150 mm 4.6 mm 5 μm Preparative Column, Standard Each
16498423 150 mm 10 mm 5 μm Preparative Column, Standard Each
16408433 150 mm 20 mm 5 μm Preparative Column, Standard Each
16418433 150 mm 25 mm 5 μm Preparative Column, Xtended Life Each
16428433 150 mm 30 mm 5 μm Preparative Column, Standard Each
16438433 150 mm 30 mm 5 μm Preparative Column, Xtended Life Each
16448433 150 mm 50 mm 5 μm Preparative Column, Standard Each
16458433 150 mm 50 mm 5 μm Preparative Column, Xtended Life Each
16468433 250 mm 2 mm 5 μm Preparative Column, Standard Each
16478433 250 mm 4.6 mm 5 μm Preparative Column, Standard Each
16488433 250 mm 10 mm 5 μm Preparative Column, Standard Each
16498433 250 mm 20 mm 5 μm Preparative Column, Standard Each
16408443 250 mm 25 mm 5 μm Preparative Column, Xtended Life Each
16418443 250 mm 30 mm 5 μm Preparative Column, Standard Each
16428443 250 mm 30 mm 5 μm Preparative Column, Xtended Life Each
16438443 250 mm 50 mm 5 μm Preparative Column, Standard Each
16448443 250 mm 50 mm 5 μm Preparative Column, Xtended Life Each
16458443 8 mm 10 mm 10 μm Guard Column Each
16468443 20 mm 10 mm 10 μm Guard Column Each
16478443 30 mm 10 mm 10 μm Guard 1/16 in. Each
16488443 30 mm 10 mm 10 μm Guard 1/8 in. Each
16498443 20 mm 20 mm 10 μm Guard Column Each
16408453 30 mm 20 mm 10 μm Guard 1/16 in. Each
16418453 30 mm 20 mm 10 μm Guard 1/8 in. Each
16428453 50 mm 10 mm 10 μm Preparative Column, Standard Each
16438453 50 mm 20 mm 10 μm Preparative Column, Standard Each
16448453 50 mm 25 mm 10 μm Preparative Column, Xtended Life Each
16458453 100 mm 10 mm 10 μm Preparative Column, Standard Each
16468453 100 mm 20 mm 10 μm Preparative Column, Standard Each
16478453 100 mm 25 mm 10 μm Preparative Column, Xtended Life Each
16488453 150 mm 10 mm 10 μm Preparative Column, Standard Each
16498453 150 mm 20 mm 10 μm Preparative Column, Standard Each
16408463 150 mm 25 mm 10 μm Preparative Column, Xtended Life Each
16418463 150 mm 30 mm 10 μm Preparative Column, Standard Each
16428463 150 mm 30 mm 10 μm Preparative Column, Xtended Life Each
16438463 150 mm 50 mm 10 μm Preparative Column, Standard Each
16448463 150 mm 50 mm 10 μm Preparative Column, Xtended Life Each
16458463 250 mm 10 mm 10 μm Preparative Column, Standard Each
16468463 250 mm 20 mm 10 μm Preparative Column, Standard Each
16478463 250 mm 25 mm 10 μm Preparative Column, Xtended Life Each
16488463 250 mm 30 mm 10 μm Preparative Column, Standard Each
16498463 250 mm 30 mm 10 μm Preparative Column, Xtended Life Each
16408473 250 mm 50 mm 10 μm Preparative Column, Standard Each
16418473 250 mm 50 mm 10 μm Preparative Column, Xtended Life Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 63 options available.
63 options
Product Code. 16498403 Supplier Thermo Scientific™ Supplier No. 41003052130
Maximize retention and loading with Hypersil PREP HS C18 prep LC reversed phase columns which offers high loading capacity and suitable for USP-L1 Applications.
Maximize retention and loading using Thermo Scientific™ Hypersil™ PREP HS C18 reversed phase preparative HPLC Columns. Our columns come in a variety of dimensions allowing you to easily scale up your analytical method, including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. These silica-based columns are packed with high surface area particles with a 100Å pore size offering C18 retention, an excellent choice for small molecule and drug purification applications.
Excellent Choice for Routine Applications
  • C18 (USP L1) selectivity - high surface area particle with 100Å pore size.
  • pH stable from 2 to 9.
  • Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
  • Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
  • Standard threaded hardware for column i.d. < 40mm.
  • Standard flange hardware for column i.d. ≥ 40mm.
  • Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
  • Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
  • Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
  • If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
  • Please contact your local sales representative for Customer Specific Orders.
  • Custom built columns are not eligible for returns.
TRUSTED_SUSTAINABILITY
Specifications
Column Format Preparative Column, Standard
Diameter (Metric) 2 mm
Length (Metric) 50 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 3 μm
pH 2 to 9
Pore Size 100 Å
Surface Area 350 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase