Learn More
Thermo Scientific™ Hypersil™ ODS C18 Columns
Shop All Thermo Scientific ProductsDescription
Use as a general purpose C18 phase for acidic and neutral analytes.
Choose Thermo Scientific™ Hypersil™ ODS C18 HPLC Columns for a general purpose C18 phase to use in a broad range of applications.
Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, the classical Hypersil phases are well established and referenced in many HPLC methods worldwide. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, we recommend the use of Thermo Scientific™ Hypersil GOLD™ columns as an alternative.
- High efficiency and proven reproducibility
- Exceptionally reliable
- Separation of a wide range of compounds including nonpolar, moderately polar and lipophilic compounds like triglycerides
Ordering Information:
Other column dimensions including preparative columns are available. Also available in 10ÎĽm. Please call Customer Service for more information.
Specifications
Specifications
Column Format | Analytical Column |
Diameter (Metric) | 3 mm |
Length (Metric) | 125 mm |
Max. Pressure | 5800 psi (400 bar) |
Particle Size | 5 ÎĽm |
pH | 2 to 8 |
Pore Size | 120 Ă… |
Surface Area | 170 m²/g |
Temperature | 60°C |
USP Type | L1 |