Learn More
Thermo Scientific™ Hypersil™ ODS C18 Columns Product Code.: 10543795
Estimated Shipment: 02-09-2025
Only null left
Add to basket
Thermo Scientific™ Hypersil™ ODS C18 Columns
Product Code. 10543795 Shop All Thermo Scientific Products
10 mm, 2.1 mm, 5 ÎĽm, Guard Cartridge, 4 Pk., Pack of 4
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
60 mm
75 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
10 mm
20 mm
Particle Size:
3 ÎĽm
5 ÎĽm
10 ÎĽm
Column Format:
Analytical Column
Guard Cartridge, 4 Pk.
Preparative Column
Preparative Column, Standard
Unit Size:
Each
Pack of 4
Product Code. 10543795
Brand: Thermo Scientific™ 30105012101
Only null left
Estimated Shipment: 02-09-2025
Add to basket
Only null left
Add to basket
Use as a general purpose C18 phase for acidic and neutral analytes.
Choose Thermo Scientific™ Hypersil™ ODS C18 HPLC Columns for a general purpose C18 phase to use in a broad range of applications.
Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, the classical Hypersil phases are well established and referenced in many HPLC methods worldwide. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, we recommend the use of Thermo Scientific™ Hypersil GOLD™ columns as an alternative.
  • High efficiency and proven reproducibility
  • Exceptionally reliable
  • Separation of a wide range of compounds including nonpolar, moderately polar and lipophilic compounds like triglycerides
TRUSTED_SUSTAINABILITY
Specifications
Column Format Guard Cartridge, 4 Pk.
Diameter (Metric) 2.1 mm
For Use With Separation of Non-Polar, Moderately Polar, Lipophilic Compounds Like Triglycerides
Length (Metric) 10 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 ÎĽm
pH 2 to 8
Pore Size 120 Ă…
Surface Area 170 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Hypersil Classical
Packing Material Silica
Carbon Load 10%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Hypersil™ ODS C18 Columns >