Learn More
Thermo Scientific™ Hypersil GOLD™ Silica HPLC Columns
Product Code. 10289124 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
100 mm
150 mm
200 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
1.9 μm
3 μm
5 μm
Unit Size:
Each
Pack of 4
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
10289124 10 mm 4 mm 3 μm Pack of 4
10311535 50 mm 2.1 mm 5 μm Each
10279644 50 mm 4.6 mm 5 μm Each
10311125 100 mm 2.1 mm 5 μm Each
10167474 150 mm 2.1 mm 5 μm Each
10437534 150 mm 4.6 mm 5 μm Each
10556955 250 mm 2.1 mm 5 μm Each
10774077 250 mm 4 mm 5 μm Each
10360835 250 mm 4.6 mm 5 μm Each
10498674 200 mm 2.1 mm 1.9 μm Each
10341165 150 mm 2.1 mm 1.9 μm Each
10065884 100 mm 2.1 mm 1.9 μm Each
10722157 50 mm 2.1 mm 1.9 μm Each
10157894 150 mm 4.6 mm 3 μm Each
10127624 150 mm 3 mm 3 μm Each
10764447 150 mm 2.1 mm 3 μm Each
10331545 100 mm 4.6 mm 3 μm Each
10066264 100 mm 2.1 mm 3 μm Each
10076744 30 mm 4.6 mm 3 μm Each
10524125 30 mm 2.1 mm 3 μm Each
10629865 10 mm 3 mm 3 μm Pack of 4
10371105 10 mm 2.1 mm 3 μm Pack of 4
10648905 10 mm 4 mm 5 μm Pack of 4
10555425 10 mm 2.1 mm 5 μm Pack of 4
24 options
Product Code. 10289124
Brand: Thermo Scientific™ 25103014001
Only null left
Add to basket
Add to basket
Efficiently separate nonpolar and moderately polar organic compounds by normal phase chromatography using Thermo Scientific™ Hypersil GOLD™ Silica LC Columns.
Achieve outstanding peak shape and sensitivity with Thermo Scientific™ Hypersil GOLD™ Silica LC Columns. The unbonded, highly pure, base-deactivated silica media efficiently separates nonpolar and moderately polar organic compounds by normal phase chromatography. Available 1.9μm particle size columns can improve the speed and efficiency of your applications.
About the Hypersil GOLD Family of HPLC Columns
  • Twelve different chemistries are available
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
TRUSTED_SUSTAINABILITY
Specifications
Column Format Guard Cartridge, 4 Pk.
Diameter (Metric) 4 mm
For Use With Uniguard Direct-Connection and Stand Alone Holder
Length (Metric) 10 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 3 μm
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
USP Type L3
Stationary Phase Silica
Product Line Hypersil GOLD
Packing Material Silica
Column Type Normal Phase
Endcapped No
Phase Normal Phase
Type Guard Cartridge