Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- HPLC Columns and Cartridges
- HPLC Columns
- Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns
Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns
Product Code. 10786153 Shop All Thermo Scientific Products| Product Code. | Length (Metric) | Diameter (Metric) | Particle Size | unitSize |
|---|---|---|---|---|
| 10786153 | 150 mm | 4.6 mm | 5 μm | Each |
| 10672854 | 50 mm | 2.1 mm | 5 μm | Each |
| 10448184 | 50 mm | 3 mm | 5 μm | Each |
| 10394173 | 50 mm | 4.6 mm | 5 μm | Each |
| 10110593 | 100 mm | 2.1 mm | 3 μm | Each |
| 10582255 | 100 mm | 2.1 mm | 5 μm | Each |
| 10354223 | 100 mm | 4.6 mm | 5 μm | Each |
| 10680136 | 100 mm | 4 mm | 3 μm | Each |
| 10763016 | 150 mm | 4 mm | 5 μm | Each |
| 11583061 | 125 mm | 4 mm | 5 μm | Each |
| 10476664 | 250 mm | 4 mm | 5 μm | Each |
| 10364653 | 250 mm | 4.6 mm | 5 μm | Each |
| 10354653 | 10 mm | 4 mm | 5 μm | Pack of 4 |
| 10662854 | 10 mm | 3 mm | 5 μm | Pack of 4 |
| 10099132 | 150 mm | 4.6 mm | 3 μm | Each |
| 10332675 | 150 mm | 3 mm | 3 μm | Each |
| 10533215 | 100 mm | 4.6 mm | 3 μm | Each |
| 10078942 | 50 mm | 4.6 mm | 3 μm | Each |
| 10600364 | 50 mm | 2.1 mm | 3 μm | Each |
| 10512065 | 10 mm | 4 mm | 3 μm | Pack of 4 |
| 10747883 | 10 mm | 3 mm | 3 μm | Pack of 4 |
| 10610764 | 10 mm | 2.1 mm | 3 μm | Pack of 4 |
| 10584285 | 150 mm | 4.6 mm | 2.4 μm | Each |
| 10457514 | 150 mm | 2.1 mm | 2.4 μm | Each |
| 10137604 | 100 mm | 4.6 mm | 2.4 μm | Each |
| 10239914 | 100 mm | 2.1 mm | 2.4 μm | Each |
| 10343105 | 50 mm | 2.1 mm | 2.4 μm | Each |
| 11969108 | 10 mm | 4 mm | 2.4 μm | Pack of 4 |
| 11919108 | 10 mm | 2.1 mm | 2.4 μm | Pack of 4 |
| 10100883 | 10 mm | 2.1 mm | 5 μm | Pack of 4 |
Descrizione
For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.
- Excellent choice for acids, bases and neutral compounds
- Less retentive than C18 for faster analyses
- Robust columns with long column lifetimes
- Very reliable for routine assays
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Specifica
Specifica
| Column Format | Analytical Column |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 150 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 130 Å |
| Surface Area | 170 m²/g |
| Temperature | 60°C |
| USP Type | L7 |
| Stationary Phase | C8 |
| Product Line | Hypersil BDS |
| Packing Material | Spherical, Fully Porous, Base-Deactivated Silica |
| Carbon Load | 7% |
| Column Type | Reversed Phase |
| Endcapped | Yes |
| Phase | Reversed Phase |
| Vedi altri risultati Mostra meno risultati |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.