Learn More
Thermo Scientific™ HyperSep™ Verify CX Cartridges
Product Code. p-4524399 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
50 mg
130 mg
200 mg
300 mg
500 mg
1000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
10 mL
Unit Size:
Pack of 100
Pack of 30
Pack of 50
Product Code. Bed Weight Column Capacity (Metric) unitSize
10796134 130 mg 1 mL Pack of 100
10130553 300 mg 3 mL Pack of 50
10300935 500 mg 3 mL Pack of 50
10557055 200 mg 6 mL Pack of 50
17287106 500 mg 6 mL Pack of 30
10058952 500 mg 6 mL Pack of 50
10629285 1000 mg 6 mL Pack of 30
11567591 200 mg 10 mL Pack of 50
11544765 130 mg 10 mL Pack of 50
12676547 200 mg 3 mL Pack of 50
11557591 50 mg 1 mL Pack of 100
11 options
Product Code. 10796134
Brand: Thermo Scientific™ 60108719
Only null left
Add to basket
Add to basket
Get improved analysis of drugs of abuse, including basic and neutral drugs, with these SPE cartridges featuring both nonpolar and ionic separation characteristics.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ Verify CX Cartridges feature both nonpolar and ionic separation chemistries for improved sample preparation in the analysis of drugs of abuse. They are used in screening for wide range of drug compounds in biological matrices, including basic and neutral compounds.
The sorbent used in HyperSep Verify CX Cartridges is based on two functional groups: reversed phase Thermo Scientific™ HyperSep C8 and a benzene sulfonic acid ion exchanger. The chemistry is co-polymerized on a rigid, purified silica gel support.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Column Capacity (Metric) 1 mL
Description 130 mg Bed Weight; 1 mL column volume
Bed Weight 130 mg
For Use With (Application) Analysis of Drugs of Abuse
For Use With (Equipment) Thermo Scientific HyperSep-96 System
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep Reversed Phase C8/Benzene Sulfonic Acid Ion Exchanger
Type Solid Phase Extraction Cartridge
Volume (Metric) 1 mL
Quantity Pack of 100