Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Shop All Thermo Scientific ProductsDescription
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
Specifications
Specifications
| Description | 24 Well Extraction Plate, 1 pk |
| For Use With (Application) | SPE Sample Processing |
| For Use With (Equipment) | Universal Vacuum Manifold |
| Product Line | HyperSep |
| Type | 24 Well Extraction Plate |