Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Product Code. 10219474 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
24 Position Extraction Plate Plugs
24 Well Extraction Plate
48 Position Extraction Plate Plugs
48 Well Extraction Plate
Unit Size:
Each
Pack of 24
Pack of 48
Product Code. Type unitSize
10219474 48 Position Extraction Plate Plugs Pack of 24
10321455 24 Position Extraction Plate Plugs Pack of 48
15277498 24 Well Extraction Plate Each
12732947 48 Well Extraction Plate Each
4 options
Product Code. 10219474
Brand: Thermo Scientific™ 60104234
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 48 Position Extraction Plate Plugs
For Use With (Application) SPE Sample Processing
For Use With (Equipment) 48 Position Well Extraction Plates, Vacuum Manifolds
Product Line HyperSep
Type 48 Position Extraction Plate Plugs