Learn More
Thermo Scientific™ HyperSep™ UniGuard Direct-Connect Online SPE Columns
Product Code. 10259224 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Hypercarb
HyperSep Quaternary Amine-Modified Polystyrene DVB
HyperSep Sulfonic Acid-Modified Polystyrene DVB
HyperSep Urea-Modified Polystyrene DVB
Diameter (Metric) Inner:
2.1 mm
3.0 mm
Unit Size:
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Diameter (Metric) Inner Unit Size
10259224 HyperSep Urea-Modified Polystyrene DVB 2.1 mm Pack of 4
10187554 HyperSep Urea-Modified Polystyrene DVB 3.0 mm Pack of 4
10259134 HyperSep Sulfonic Acid-Modified Polystyrene DVB 2.1 mm Pack of 4
10783487 HyperSep Sulfonic Acid-Modified Polystyrene DVB 3.0 mm Pack of 4
10005984 HyperSep Quaternary Amine-Modified Polystyrene DVB 2.1 mm Pack of 4
10772147 HyperSep Quaternary Amine-Modified Polystyrene DVB 3.0 mm Pack of 4
11584785 HyperSep Hypercarb 2.1 mm Pack of 4
10015794 HyperSep Hypercarb 3.0 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10259224 Supplier Thermo Scientific™ Supplier No. 60311201
Estimated Shipment: 06-10-2026
Clean-up and concentrate target analytes in minutes with these direct-connection SPE cartridges, offering quick, easy connection via a reusable drop-in cartridge holder.
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Column offer quick, easy connection by means of a reusable drop-in guard cartridge holder.
HyperSep UNIGUARD Direct-Connect Online SPE Cartridges attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
TRUSTED_SUSTAINABILITY
Specifications
Description Hypersep Retain PEP; 2.1 mm ID
For Use With (Application) Retention of Polar and Nonpolar Analytes
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Online Solid Phase Extraction Cartridge
Quantity Pack of 4
Diameter (Metric) Inner 2.1 mm