Learn More
Thermo Scientific™ HyperSep™ UniGuard Direct-Connect Online SPE Columns
Product Code. 10259134 Shop All Thermo Scientific Products
Click to view available options
Sorbent:
HyperSep Hypercarb
HyperSep Quaternary Amine-Modified Polystyrene DVB
HyperSep Sulfonic Acid-Modified Polystyrene DVB
HyperSep Urea-Modified Polystyrene DVB
Diameter (Metric) Inner:
2.1 mm
3.0 mm
Unit Size:
Pack of 4
Product Code. 10259134
Brand: Thermo Scientific™ 60311301
Only null left
Add to basket
Add to basket
Clean-up and concentrate target analytes in minutes with these direct-connection SPE cartridges, offering quick, easy connection via a reusable drop-in cartridge holder.
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep UniGuard Direct-Connect Online SPE Column offer quick, easy connection by means of a reusable drop-in guard cartridge holder.
HyperSep UNIGUARD Direct-Connect Online SPE Cartridges attach directly to the switching valve, just like an HPLC column. They are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
TRUSTED_SUSTAINABILITY
Specifications
Description HyperSep Retain-CX; 2.1 mm ID
For Use With (Application) Retention of Basic Analytes
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Sulfonic Acid-Modified Polystyrene DVB
Type Online Solid Phase Extraction Cartridge
Quantity Pack of 4
Diameter (Metric) Inner 2.1 mm
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis