Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 10-200μL
Product Code. 10516015 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
BioBasic C18
BioBasic C4
BioBasic C8
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Unit Size:
Pack of 96
Product Code. Sorbent unitSize
10516015 HyperSep Zirconium Dioxide Pack of 96
10191213 BioBasic C18 Pack of 96
10566375 BioBasic C8 Pack of 96
10344083 BioBasic C4 Pack of 96
10775174 HyperSep Hypercarb and C18 (Mix Mode) Pack of 96
10465363 HyperSep HILIC Pack of 96
10055824 HyperSep Trypsin Pack of 96
10652094 HyperSep Titanium Dioxide Pack of 96
10223603 HyperSep Hypercarb Pack of 96
9 options
Product Code. 10516015
Brand: Thermo Scientific™ 60109218
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
Mass spectrometry
Desalting
Protein purification
MALDI
Electrophoresis
HPCE
HPLC
CEC
Tips are available in 1-10µL capacity and 10-200µL capacity.
TRUSTED_SUSTAINABILITY
Specifications
Description Zirconium Dioxide; 10 to 200 uL Volume
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Product Line HyperSep
Sorbent HyperSep Zirconium Dioxide
Type SPE Tips
Volume (Metric) 10 to 200 μL