Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL
Shop All Thermo Scientific ProductsDescription
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
- [bullet] Filter-free design to eliminate dead volume
- [bullet] No contamination from the supporting matrix
- [bullet] Available in many popular chemistries
- [bullet] Mass spectrometry
- [bullet] Desalting
- [bullet] Protein purification
- [bullet] MALDI
- [bullet] Electrophoresis
- [bullet] HPCE
- [bullet] HPLC
- [bullet] CEC
Spécification
Spécification
| Description | C18 plus Hypercarb; 10-200 uL Vol. |
| For Use With (Application) | Microscale Solid Phase Extraction |
| For Use With (Equipment) | Micropipettes |
| Product Line | HyperSep |
| Sorbent | HyperSep Hypercarb and C18 (Mix Mode) |
| Type | SpinTip |
| Volume (Metric) | 10 to 200 μL |