Description
Provide a fast and effective sample preparation technique for removal of phospholipids from biological matrices with Thermo Scientific™ HyperSep™ SLE (Solid supported Liquid/Liquid Extraction).
HyperSep SLE cartridges (pH 7) deals with sample preparation of biological matrices via a simple, efficient and an effective protcol. The focus being the removal of interfering phospholipids which cause a number of issues for end point analysis.
- Simple, easy-to-use protocol
- Removal of phospholipids
- Greater reproducability and recoveries compared to LLE
Specifications
Specifications
| Description | 500 mg/6 mL, Pk. of 30, pH7 |
| Bed Weight | 500 mg |
| For Use With (Application) | Removal of Phospholipids From Biological Matrices |
| Product Line | HyperSep |
| Sorbent | HyperSep Special Treated Diatomite |
| Type | Solid Phase Extraction Cartridge |
| Volume (Metric) | 6 mL |
| Quantity | Pack of 30 |