Learn More
Thermo Scientific™ HyperSep™ Servo+ Total SPE Cartridges and Well Plates
Product Code. 11352273 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
1 g 25 mL SPE Column 20 Pk
150 mg 6 mL SPE Column 30 Pk
200 mg 6 mL SPE Column 30 Pk
200 mg/3 mL SPE Column 50 Pk
500 mg 6 mL SPE Column 30 Pk
500 mg/3 mL SPE Column 50 Pk
60 mg 6 mL SPE Column 30 Pk
60 mg/3 mL SPE Column 50 Pk
96-well plate 10 mg 1 Pk
96-well plate 30 mg 1 Pk
96-well plate 5 mg 1 Pk
Bed Weight:
5 mg
10 mg
30 mg
60 mg
150 mg
200 mg
500 mg
1 g
Unit Size:
Each
Pack of 20
Pack of 30
Pack of 50
Volume (Metric):
3 mL
6 mL
25 mL
Product Code. Description Bed Weight unitSize Volume (Metric)
11352273 96-well plate 5 mg 1 Pk 5 mg Each -
11372273 96-well plate 10 mg 1 Pk 10 mg Each -
11382273 60 mg/3 mL SPE Column 50 Pk 60 mg Pack of 50 3 mL
11392273 96-well plate 30 mg 1 Pk 30 mg Each -
11302283 200 mg/3 mL SPE Column 50 Pk 200 mg Pack of 50 3 mL
11312283 500 mg/3 mL SPE Column 50 Pk 500 mg Pack of 50 3 mL
11322283 500 mg 6 mL SPE Column 30 Pk 500 mg Pack of 30 6 mL
11342283 60 mg 6 mL SPE Column 30 Pk 60 mg Pack of 30 6 mL
11352283 150 mg 6 mL SPE Column 30 Pk 150 mg Pack of 30 6 mL
11362283 200 mg 6 mL SPE Column 30 Pk 200 mg Pack of 30 6 mL
11372283 1 g 25 mL SPE Column 20 Pk 1 g Pack of 20 25 mL
11 options
Product Code. 11352273
Brand: Thermo Scientific™ 60110101P
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Screening for unknown components when analyzing drugs of abuse? These SPE cartridges and plates offer a robust, ready-to-use solution.
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo+ products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo+ Total Cartridges and Well Plates offer a robust, ready-to-use solution for screening of a wide range of unknown components.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 96-well plate 5 mg 1 Pk
Bed Weight 5 mg
For Use With (Application) Analysis of Drugs of Abuse
Product Line HyperSep
Sorbent HyperSep Servo+ Total
Type 96-Well Plate