Learn More
Thermo Scientific™ HyperSep™ Retain PEP Cartridges
Product Code. p-4524366 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
30 mg
60 mg
100 mg
150 mg
200 mg
300 mg
500 mg
600 mg
1000 mg
2000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
25 mL
Unit Size:
Pack of 100
Pack of 20
Pack of 30
Pack of 50
Product Code. Bed Weight Column Capacity (Metric) unitSize
10595895 30 mg 1 mL Pack of 100
15974417 600 mg 6 mL Pack of 50
10259564 30 mg 3 mL Pack of 50
15984417 300 mg 3 mL Pack of 100
10505905 60 mg 3 mL Pack of 50
10448104 200 mg 3 mL Pack of 50
10351215 500 mg 3 mL Pack of 50
10618725 500 mg 6 mL Pack of 30
10015894 100 mg 6 mL Pack of 30
10269564 60 mg 6 mL Pack of 30
10279954 150 mg 6 mL Pack of 30
10565245 200 mg 6 mL Pack of 30
11504745 2000 mg 25 mL Pack of 20
11514745 1000 mg 25 mL Pack of 20
13406936 100 mg 3 mL Pack of 50
13436936 1000 mg 6 mL Pack of 30
16 options
Product Code. 10595895
Brand: Thermo Scientific™ 60107201
Only null left
Add to basket
Add to basket
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features:
Ideal for large samples, Hypersep Retain SPE cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • Available in a wide range of chemistries and bed weights
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifications
Column Capacity (Metric) 1 mL
Description 30 mg bed weight; 1 mL column volume
Bed Weight 30 mg
For Use With (Application) Retention of Polar and Nonpolar Analytes
For Use With (Equipment) Thermo Scientific HyperSep-96 System
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Solid Phase Extraction Cartridge
Volume (Metric) 1 mL
Quantity Pack of 100