Learn More
Thermo Scientific™ HyperSep™ Retain PEP Cartridges
Product Code. 15984417 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
30 mg
60 mg
100 mg
150 mg
200 mg
300 mg
500 mg
600 mg
1000 mg
2000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
25 mL
Unit Size:
Pack of 100
Pack of 20
Pack of 30
Pack of 50
Product Code. Bed Weight Column Capacity (Metric) unitSize
15984417 300 mg 3 mL Pack of 100
10595895 30 mg 1 mL Pack of 100
15974417 600 mg 6 mL Pack of 50
10259564 30 mg 3 mL Pack of 50
10505905 60 mg 3 mL Pack of 50
10448104 200 mg 3 mL Pack of 50
10351215 500 mg 3 mL Pack of 50
10618725 500 mg 6 mL Pack of 30
10015894 100 mg 6 mL Pack of 30
10269564 60 mg 6 mL Pack of 30
10279954 150 mg 6 mL Pack of 30
10565245 200 mg 6 mL Pack of 30
11504745 2000 mg 25 mL Pack of 20
11514745 1000 mg 25 mL Pack of 20
13406936 100 mg 3 mL Pack of 50
13436936 1000 mg 6 mL Pack of 30
16 options
Product Code. 15984417
Brand: Thermo Scientific™ 60107226
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features:
Ideal for large samples, Hypersep Retain cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • Available in a wide range of chemistries and bed weights
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifications
Column Capacity (Metric) 3 mL
Description 300 mg bed weight; 3 mL column volume
Bed Weight 300 mg
For Use With (Application) Retention of Polar and Nonpolar Analytes
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Solid Phase Extraction Cartridge
Volume (Metric) 3 mL