Learn More
Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells
Product Code. 10661016 Shop All Thermo Scientific Products
Click to view available options
Sorbent:
HyperSep Retain PEP
missing translation for 'unitSize'
Each
Product Code. 10661016
missing translation for 'mfr': Thermo Scientific™ 60303208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifications
Description 96-Well Plate; Bed weight 60 mg
For Use With (Application) Screening of Drugs, Metabolites, Peptides in Biological Samples
Particle Size 30 to 50 μm
Product Line HyperSep
Sorbent HyperSep Retain PEP
Type 96-well Plate