Learn More
Thermo Scientific™ HyperSep™ Javelin Direct-Connect Online SPE Columns
Product Code. 10771957 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Hypercarb
HyperSep Retain AX
HyperSep Retain CX
HyperSep Retain PEP
Diameter (Metric) Inner:
2.1 mm
3.0 mm
Unit Size:
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Diameter (Metric) Inner Unit Size
10771957 HyperSep Retain PEP 3.0 mm Pack of 4
10128044 HyperSep Retain PEP 2.1 mm Pack of 4
10594275 HyperSep Retain CX 2.1 mm Pack of 4
10504285 HyperSep Retain CX 3.0 mm Pack of 4
10504855 HyperSep Retain AX 2.1 mm Pack of 4
10167694 HyperSep Retain AX 3.0 mm Pack of 4
10565225 HyperSep Hypercarb 2.1 mm Pack of 4
10239314 HyperSep Hypercarb 3.0 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10771957 Supplier Thermo Scientific™ Supplier No. 60310202
Estimated Shipment: 27-11-2026
Clean-up and concentrate target analytes in minutes with these direct-connection SPE guard columns, featuring fingertight design for convenience and efficiency.
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep Javelin Direct-Connect Online SPE Columns feature a unique fingertight design for convenience and efficiency.
HyperSep Javelin Direct-Connect Online SPE Columns are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
TRUSTED_SUSTAINABILITY
Specifications
Description HyperSep Retain PEP; 3.0 mm ID
For Use With (Application) Online Solid Phase Extraction
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Retain PEP
Type Online Solid Phase Extraction Column
Diameter (Metric) Inner 3.0 mm