Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Adaptors Safety Tray
Product Code. 10007654 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Manifold Safety Tray, 1 pk
Unit Size:
Each
Product Code. Description unitSize
10007654 Manifold Safety Tray, 1 pk Each
1 options
Product Code. 10007654
Brand: Thermo Scientific™ 60104260
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use the safety tray as the bottom of the glass block vacuum manifold to contain spills.
Use the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Safety Tray as the bottom of the manifold and to contain spills. The HyperSep Glass Block Vacuum Manifold enables solid phase extraction sample preparation using 16 or 24 SPE cartridges.
TRUSTED_SUSTAINABILITY
Specifications
Description Manifold Safety Tray, 1 pk
For Use With (Application) SPE Sample Preparation
For Use With (Equipment) Glass Block Vacuum Manifold Adaptors
Product Line HyperSep
Type Manifold Safety Tray