Learn More
Thermo Scientific™ HyperSep™ C18 Cartridges
Product Code. 15982217 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
25 mg
50 mg
100 mg
200 mg
500 mg
1000 mg
2000 mg
5000 mg
10 g
Column Capacity (Metric):
1 mL
3 mL
6 mL
10 mL
15 mL
25 mL
75 mL
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 30
Pack of 50
Product Code. Bed Weight Column Capacity (Metric) unitSize
15982217 500 mg 6 mL Pack of 30
10344653 1000 mg 6 mL Pack of 30
10571295 100 mg 1 mL Pack of 100
10631324 200 mg 3 mL Pack of 50
10434963 500 mg 3 mL Pack of 50
10098942 500 mg 6 mL Pack of 30
10610436 50 mg 1 mL Pack of 100
10523364 2000 mg 15 mL Pack of 20
10166203 5000 mg 25 mL Pack of 20
10434445 10 g 75 mL Pack of 10
11514765 100 mg 3 mL Pack of 50
11830493 25 mg 1 mL Pack of 100
13476936 500 mg 10 mL Pack of 50
13 options
Product Code. 15982217
Brand: Thermo Scientific™ 60108305P
Only null left
Add to basket
Add to basket
Get improved retention for most nonpolar compounds and organic analytes in aqueous matrices with these hydrophobic reversed phase C18 cartridges.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications. Thermo Scientific™ HyperSep™ C18 Cartridges feature a highly retentive alkyl-bonded phase for nonpolar to moderately polar compounds. The hydrophobic reversed phase material is retentive for most nonpolar compounds, and retains most organic analytes from aqueous matrices.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Column Capacity (Metric) 6 mL
Description HyperSep C18 Cartridge
Bed Weight 500 mg
For Use With (Application) Extraction of Nonpolar to Moderately Polar Compounds
For Use With (Equipment) Thermo Scientific HyperSep-96 System
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep C18
Type Solid Phase Extraction Cartridge
Volume (Metric) 6 mL
Quantity Pack of 30