Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Manifold, Gauge and Waste Base
Product Code. 10384313 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Universal Vacuum Manifold
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10384313 Universal Vacuum Manifold Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10384313 Supplier Thermo Scientific™ Supplier No. 60104231
Estimated Shipment: 18-12-2026
Use this simple vacuum manifold for solid phase extraction (SPE) in 96 well plates.
Order the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold, Gauge and Waste Base as a replacement spare part for your HyperSep Universal Vacuum Manifold. This manifold processes samples in solid phase extraction SPE cartridges or 96 well plate.
TRUSTED_SUSTAINABILITY
Specifications
Description Universal Vacuum Manifold, Gauge and Waste Base
For Use With (Application) SPE
For Use With (Equipment) Universal Vacuum Manifold
Product Line HyperSep
Type Gauge and Waste Base