Learn More
Safety First Aid HypaSoothe Emergency Burn Gel, Medium
Product Code. 31759443
Change view
Click to view available options
Size:
Medium
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31759443 Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31759443 Supplier Safety First Aid Supplier No. D8164
  • ONLINE EXCLUSIVE PRICE
    31.27 NOK
    28.65 NOK / Pack of 1
    Save 2.62 NOK
    8% Off
 Restricted Item
Estimated Shipment: 04-12-2026
The gel is a sterile wound hydrogel that facilitates moist wound healing for minor burns. The gel re-hydrates necrotic tissue while loosening and absorbing slough and exudate.
  • The gel is suitable for use on 1st and 2nd degree burns.
  • It soothes burns and reduces the likelihood of scarring by creating an ideal wound healing environment.
  • High cohesion means the gel is easily removed, even after the absorption of debris and exudate.
  • Supplied in a 20 pack of 6 g sachets.
  • Usage: Cool the burn with water. Gently dry the skin around the wound and apply the gel Cover the wound with a secondary dressing. Caution: For external use only. Do not reuse gel that has already been applied. Do not use if packaging is broken.
  • In the case of an allergic reaction, stop usage and consult a healthcare professional
Specifications
Material Cloth
Product Type Burn Dressing
Sterility Non-sterile
Quantity 50 g
Size Medium
Packaging Type Bottle