Learn More
Safety First Aid HypaCover Gauze Swabs
Product Code. 31759444
Change view
Click to view available options
Dimensions (L x W):
7.5 x 7.5 cm
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
31759444 7.5 x 7.5 cm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31759444 Supplier Safety First Aid Supplier No. D7871PK20
  • ONLINE EXCLUSIVE PRICE
    70.20 NOK
    64.32 NOK / Pack of 20
    Save 5.88 NOK
    8% Off
 Restricted Item
Estimated Shipment: 04-12-2026
The Sterile Gauze Swabs are protective cotton swabs ideal for cushioning, cleaning and protecting wounds.
  • Sterile and non-sterile Gauze swabs from Safety First Aid.
  • Use these highly absorbent cotton swabs to clean wounds and prepare them for dressing.
  • Non-sterile swabs are available in three different sizes.
  • The highly absorbent cotton gauze has a soft yet slightly abrasive texture, and allows blood and other fluids to pass through and be absorbed.
  • Delivered in 20 packs of 5 swabs for a total of 10.
Specifications
Material Gauze
Product Type Gauze Bandage
Sterility Sterile
Quantity 100 Pk.
Size Large
Dimensions (L x W) 7.5 x 7.5 cm