Learn More
Safety First Aid HypaCover Alcohol-Free Moist Wipes, Sterile
Product Code. 31759429
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31759429 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31759429 Supplier Safety First Aid Supplier No. D5201
  • ONLINE EXCLUSIVE PRICE
    64.14 NOK
    58.79 NOK / Pack of 1
    Save 5.35 NOK
    8% Off
 Restricted Item
Estimated Shipment: 09-12-2026
The HypaCover Sterile Moist Wipes, Alcohol-Free (100 Pk.) are premium wipes designed for optimal wound cleansing and skin hygiene. Saturated with a gentle, aqueous solution, these individually wrapped sterile wipes are safe for use directly on open wounds, without stinging or irritation.
  • Key Benefits:
    • Ultra gentle, alcohol free cleansing helps maintain skin integrity and supports faster wound healing, ideal for sensitive or eczema prone skin.
    • Individually sealed pouches preserve sterility and moisture, so each wipe is fresh and effective.
    • Versatile for multiple settings: essential in hospitals, schools, labs, food prep zones, sports events, labs and household first aid kits.
    • Perfect for travel and emergency kits - lightweight, compact 10 x 12 cm size, essential for on the go care.
    • Nearly twice the size of many other brands on the market, offering more coverage for effective, thorough cleaning.
    • Their larger size means fewer wipes are needed to clean around wounds-reducing waste, saving time, and providing better value.
  • Recommended Use Cases:
    • Emergency wound cleaning: syringes, cuts, scrapes - clean before dressing.
    • Field sports schools: convenient hygiene when accidents occur outdoors.
    • Workplace first aid: meets regulatory standards and supports staff welfare.
    • Sensitive skin applications: no burning sensation; prevents alcohol irritation.
  • What’s Inside:
    • 100 x sterile, alcohol free wipes, individually wrapped.
    • Wipe size: 10 x 12 cm (unfolded), compact when sealed.
  • Why Customers Choose HypaCover:
    • Promotes clean, infection free healing without alcohol pain.
    • Easily meets legal and safety compliance for schools and workplaces.
    • Budget friendly bulk option, great value for frequent use.
    • Compact packaging supports portability and emergency readiness.
Specifications
Product Type Skin Cleansing Wipe
Color White
Form Wipe