Learn More
Safety First Aid HypaClens Sterile Eye Wash Pods
Product Code. 31759384
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
31759384 Sterile Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31759384 Supplier Safety First Aid Supplier No. E401APK25
  • ONLINE EXCLUSIVE PRICE
    65.57 NOK
    60.07 NOK / Pack of 25
    Save 5.50 NOK
    8% Off
 Restricted Item
Estimated Shipment: 16-12-2026
The eyewash pods gently wash the eye and provide moisture-replenishing relief for eyes that have been contaminated with particles or liquids. Their 0.9% sterile saline solution is ideal for eye irrigation.
  • HypaClens Sterile Eyewash Pods provide eye wash in a compact and easily portable size.
  • These 20 mL pods are ideal for inclusion in first aid kits where 250 mL or 500 mL bottles would be impractical.
  • They can be used to cleanse the eye of dust, dirt, small insects and other irritants.
  • For minor, non-emergency use, these pods are the perfect alternative to using a larger bottle, as eyewash bottles are single-use and cannot be resealed.
  • However, the pods are also ideal in the workplace as a portable, rapid-response item for responding to eye contamination at the scene of an incident.
Specifications
Product Type Eyewash Solution
Quantity 25 x 20 mL