Learn More
Safety First Aid HypaClens Emergency Eye Wash Dispenser, 10 x 20 mL Pods
Product Code. 31759396
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
31759396 Sterile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31759396 Supplier Safety First Aid Supplier No. E4201
  • ONLINE EXCLUSIVE PRICE
    56.08 NOK
    51.39 NOK / Pack of 1
    Save 4.69 NOK
    8% Off
Estimated Shipment: 04-12-2026
HypaClens Sterile Eyewash Pods gently wash the eye and provide sensitive and moisture replenishing relief for eyes that have been contaminated with particles or liquid.
  • HypaClens Sterile Eyewash Pods contain 0.9% sterile saline solution that is ideal for irrigating eyes and general wound washing applications.
  • The station can be refilled when required.
Specifications
Product Type Eyewash Solution
Sterility Sterile
Volume (Metric) 10 x 20 mL