Learn More
Hyoscyamine, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Hyoscyamine |
| CAS | 101-31-5 |
| Analyte or Component Names | Hyoscyamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H23 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1αH,5αH-Tropan-3α-ol, (-)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, [3(S)-endo]-, Tropic acid, 1αH,5αH-tropan-3α-yl ester, (-)- (8CI), (-)-Atropine, (-)-Hyoscyamine, (S)-(-)-Hyoscyamine, (S)-Atropine, 1αH,5αH-Tropan-3α-yl (-)-tropate, Cystospaz, Daturine, Duboisine, Hyoscyamine, L-Hyoscyamin, L-Hyoscyamine, l-Atropine, l-Hyoscyamine, l-Tropine tropate, Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, (αS)-, Hyoscine Hydrobromide Impurity A, Hyoscine Impurity A |
| InChI Formula | InChI=1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16-/m1/s1 |
| Solution Type | Neat |
| SMILES | CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](CO)c3ccccc3 |
| Show More |