Learn More
Hygromycin B, 96.34%, MP Biomedicals™
Product Code. 15284578 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 kU
Unit Size:
250ku
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15284578 250 kU 250ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15284578 Supplier MP Biomedicals Supplier No. 0215751383
  • ONLINE EXCLUSIVE PRICE
    1835.00 NOK
    1746.31 NOK / 250ku
    Save 88.69 NOK
    4% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Antibacterial and antifungal agent that inhibits protein synthesis by interfering with ribosome translocation.
Specifications
Type Hygromycin B
Physical Form Liquid
CAS 31282-04-9, 7732-18-5
Color Brown to Red
Quantity 250 kU
Assay Percent Range 0.9634
Molecular Formula C20H37N3O13
Synonym Antihelmycin, Hygromix-8
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
Molecular Weight (g/mol) 527.524
PubChem CID 134129613
Formula Weight 527.5
Percent Purity ≥80%
Grade Not Available
Chemical Name or Material Hygromycin B